Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAQTPAFDK PKVELHVHLD GSIKPETILY YGRRRGIALP ANTAEGLLNV IGMDKPLTLP DFLAKFDYYM PAIAGCREAI KRIAYEFVEM KAKEGVVYVE VRYSPHLLAN SKVEPIPWNQ AEGDLTPDEV VALVGQGLQE GERDFGVKAR SILCCMRHQP NWSPKVVELC KKYQQQTVVA IDLAGDETIP GSSLLPGHVQ AYQEAVKSGI HRTVHAGEVG SAEVVKEAVD ILKTERLGHG YHTLEDQALY NRLRQENMHF EICPWSSYLT GAWKPDTEHA VIRLKNDQAN YSLNTDDPLI FKSTLDTDYQ MTKRDMGFTE EEFKRLNINA AKSSFLPEDE KRELLDLLYK AYGMPPSASA GQNL (1-363 aa encoded by BC040226)
Catalyzes the hydrolytic deamination of adenosine and 2-deoxyadenosine. Plays an important role in purine metabolism and in adenosine homeostasis. Modulates signaling by extracellular adenosine, and so contributes indirectly to cellular signaling events. May act as a positive regulator of T-cell coactivation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenosine aminohydrolase; Adenosine deaminase; RP11-61L14.5
Gene Aliases: ADA; ADA1
UniProt ID: (Human) P00813, (Mouse) P03958
Entrez Gene ID: (Human) 100, (Mouse) 11486
Molecular Function: deaminase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support