Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HRLCCVSCIK KASGPNPGPD PGPTSLPPFS TPGSPLPGPQ DPADAAEPPG KPTGSEDHQH GRATQLPGAL DTSSPGTQHP FAPETPIPGA SWSISPTTPG GLPWGWTQTP TPVPEDKGQP GEDLRHPGTS LPAASPVT
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. This gene is highly similar to two family members, ADAMTS2 and ADAMTS3, in its sequence and gene structure, and the encoded protein shares the aminoprocollagen peptidase activity with the protein products encoded by ADAMTS2 and ADAMTS3. Various transcript variants of this gene have been identified. They result from the use of two different promoters and transcription initiation sites as well as alternative splicing sites. The full length nature of some transcripts has not been defined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 14; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 14; ADAM; ADAM-TS 14; ADAM-TS14; ADAMs; ATS14; FLJ32820; metalloendopeptidases
Gene Aliases: ADAMTS14
UniProt ID: (Human) Q8WXS8
Entrez Gene ID: (Human) 140766
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support