Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: ESCSKRSSTL PPPYLLEAAE THDDVISNPS DLPRSLVMPT SLVPYHSETP AKKMSLSSIS SVGGPNAYAA FRPNSKPDGA N
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene is the major procollagen II N-propeptidase. A deficiency of this protein may be responsible for dermatosparaxis, a genetic defect of connective tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 3; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 3; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif, 3; ADAM-TS 3; ADAM-TS3; ADAMs; ADAMTS-3; metalloendopeptidases; PC II-NP; Procollagen II amino propeptide-processing enzyme; Procollagen II N-proteinase; zinc metalloendopeptidase
Gene Aliases: ADAMTS-4; ADAMTS3; KIAA0366
UniProt ID: (Human) O15072
Entrez Gene ID: (Human) 9508
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support