Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Antibody detects endogenous levels of total ADCK1.
ADCK1 is a protein coding gene. The function of this protein is not yet clear. It is not known if it has protein kinase activity and what type of substrate it would phosphorylate (Ser, Thr or Tyr).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AarF domain-containing protein kinase 1; Nrbf1; uncharacterized aarF domain-containing protein kinase 1
Gene Aliases: 2610005A10Rik; ADCK1
UniProt ID: (Human) Q86TW2, (Mouse) Q9D0L4
Entrez Gene ID: (Human) 57143, (Mouse) 72113
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support