Immunogen sequence: MRAHAAALG VQDYLLFAEM LMQRPVRLGQ LWGSHLLSRE EAAYMVDMAR ERFEAVMAVL RELPRPMLLV LRNINTVRAI NVALGAPVDR YFLMAKRAVR GWSRLAGATY RGVYGTSLLR HAKVVWEMLK FEVALRLETL AMRLTALLAR ALVHLSLVPP AEELYQYLET (1-169 aa encoded by BC031570)
ADCK5 is member of the AarF domain containing kinase family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Uncharacterized aarF domain-containing protein kinase 5
Gene Aliases: A230108P17; ADCK5
UniProt ID: (Human) Q3MIX3, (Mouse) Q80V03
Entrez Gene ID: (Human) 203054, (Rat) 362943, (Mouse) 268822
Molecular Function:
ATP-binding cassette (ABC) transporter
primary active transporter
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support