Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: AEVSQRQAF QETRGYIQAR LHLQHENRQQ ERLLLSVLPQ HVAMEMKEDI NTKKEDMMFH KIYIQKHDNV SILFADIEGF TSLASQCTAQ ELVMTLNELF ARFDKLAAEN HCLRIKILGD CYYCVSGLPE ARADHAHCCV EMGVDMIEAI SLVREVTGVN VNMRVGIHSG RVHCGVLGLR KWQFDVWSND VTLANHMEAG GRAGRIHITR ATLQYLNGDY EVEPGRGGER NAYLKEQHIE TFLILGASQK RKEEKAMLAK LQRTRANSME GLMPRWVPDR AFSRTKDSKA FRQMGIDDSS KDNRGTQDAL NPEDEVDEFL SRAIDARSID QLRKDHVRRF LLTFQRE (310-655 aa encoded by BC064923 )
This gene encodes a member of the adenylyl cyclase family of proteins, which are required for the synthesis of cyclic AMP. All members of this family have an intracellular N-terminus, a tandem repeat of six transmembrane domains separated by a cytoplasmic loop, and a C-terminal cytoplasmic domain. The two cytoplasmic regions bind ATP and form the catalytic core of the protein. Adenylyl cyclases are important effectors of transmembrane signaling pathways and are regulated by the activity of G protein coupled receptor signaling. This protein belongs to a small subclass of adenylyl cyclase proteins that are functionally related and are inhibited by protein kinase A, calcium ions and nitric oxide. A mutation in this gene is associated with arthrogryposis multiplex congenita.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AC6; ACVI; Adenylate cyclase type 6; Adenylate cyclase type VI; Adenylyl cyclase 6; ATP pyrophosphate-lyase 6; Ca(2+)-inhibitable adenylyl cyclase; DKFZp779F075
Gene Aliases: AC6; ACVI; ADCY6; ADCYB; KIAA0422; LCCS8; mKIAA0422
UniProt ID: (Human) O43306, (Rat) Q03343
Entrez Gene ID: (Human) 112, (Rat) 25289, (Mouse) 11512
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support