Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MNGDSRAAVV TSPPPTTAPH KERYFDRVDE NNPEYLRERN MAPDLRQDFN MMEQKKRVSM ILQSPAFCEE LESMIQEQFK KGKNPTGLLA LQQIADFMTT NVPNVYPAAP QGGMAALNMS LGMVTPVNDL RGSDSIAYDK GEKLLRCKLA AFYRLADLFG WSQLIYNHIT TRVNSEQEHF LIVPFGLLYS EVTASSLVKI NLQGDIVDRG STNLGVNQAG FTLHSAIYAA RPDVKCVVHI HTPAGAAVSA MKCGLLPISP EALSLGEVAY HDYHGILVDE EEKVLIQKNL GPKSKVLILR NHGLVSVGES VEEAFYYIHN LVVACEIQVR TLASAGGPDN LVLLNPEKYK AKSRSPGSPV GEGTGSPPKW QIGEQEFEAL MRMLDNLGYR TGYPYRYPAL REKSKKYSDV EVPASVTGYS FASDGDSGTC SPLRHSFQKQ QREKTRWLNS GRGDEASEEG QNGSSPKSKT KWTKEDGHRT STSAVPNLFV PLNTNPKEVQ EMRNKIREQN LQDIKTAGPQ SQVLCGVVMD RSLVQDAPLS DCTETIEGLE LTEQTFSPAK SLSFRKGELV TASKAIIEKE YQPHVIVSTT GPNPFTTLTD RELEEYRREV ERKQKGSEEN LDEAREQKEK SPPDQPAVPH PPPSTPIKLE EGDGCAREYL LP
Adducins are a family of cytoskeleton proteins encoded by three genes (alpha, beta, gamma). Adducin is a heterodimeric protein that consists of related subunits, which are produced from distinct genes but share a similar structure. Alpha- and beta-adducin include a protease-resistant N-terminal region and a protease-sensitive, hydrophilic C-terminal region. Alpha- and gamma-adducins are ubiquitously expressed. In contrast, beta-adducin is expressed at high levels in brain and hematopoietic tissues. Adducin binds with high affinity to Ca(2+)/calmodulin and is a substrate for protein kinases A and C. Alternative splicing results in multiple variants encoding distinct isoforms; however, not all variants have been fully described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADD2; ADDB; adducin 1 (alpha); Adducin beta; Adducin-1; Adducin-alpha; Adducin-beta; Adducin1; Alpha adducin; Alpha-adducin; beta Adducin; erythrocyte adducin alpha subunit; Erythrocyte adducin subunit alpha
Gene Aliases: ADD1; ADDA
UniProt ID: (Human) P35611
Entrez Gene ID: (Human) 118
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support