Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Additional Information: This antibody is expected to recognise the alpha (ADH1A, NP_ 000658.1), the beta (ADH1B, NP_000659.2) and gamma (ADH1C, NP_000660.1) polypeptide variants of human alcohol dehydrogenase.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
Alcohol dehydrogenase,consisting of several homo- and heterodimers of alpha, beta, and gamma subunits, exhibits high activity for ethanol oxidation and plays a major role in ethanol catabolism.Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. Three genes encoding alpha, beta and gamma subunits are tandemly organized in a genomic segment as a gene cluster.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADH, alpha subunit; ADH, beta subunit; ADH, gamma subunit; alcohol dehydrogenase 1 (class I), alpha polypeptide; Alcohol dehydrogenase 1A; Alcohol dehydrogenase 1B; Alcohol dehydrogenase 1C; alcohol dehydrogenase 2 (class I), beta polypeptide; alcohol dehydrogenase 3 (class I), gamma polypeptide; Alcohol dehydrogenase subunit alpha; Alcohol dehydrogenase subunit beta; Alcohol dehydrogenase subunit gamma; aldehyde reductase; All-trans-retinol dehydrogenase [NAD(+)] ADH1B; epididymis secretory protein Li 117
Gene Aliases: ADH1; ADH1A; ADH1B; ADH1C; ADH2; ADH3; HEL-S-117
UniProt ID: (Human) P07327, (Human) P00325, (Human) Q4PJ18
Entrez Gene ID: (Human) 124, (Human) 125, (Human) 126
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support