Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGTKGKVIK CKAAIAWEAG KPLCIEEVEV APPKAHEVRI QIIATSLCHT DATVIDSKFE GLAFPVIVGH EAAGIVESIG PGVTNVKPGD KVIPLYAPLC RKCKFCLSPL TNLCGKISNL KSPASDQQLM EDKTSRFTCK GKPVYHFFGT STFSQYTVVS DINLAKIDDD ANLERVCLLG CGFSTGYGAA INNAKVTPGS TCAVFGLGGV GLSAVMGCKA AGASRIIGID INSEKFVKAK ALGATDCLNP RDLHKPIQEV IIELTKGGVD FALDCAGGSE TMKAALDCTT AGWGSCTFIG VAAGSKGLTV FPEELIIGRT INGTFFGGWK SVDSIPKLVT DYKNKKFNLD ALVTHTLPFD KISEAFDLMN QGKSIRTILI F (1-380 aa encoded by BC022319)
This gene encodes class II alcohol dehydrogenase 4 pi subunit, which is a member of the alcohol dehydrogenase family. Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. Class II alcohol dehydrogenase is a homodimer composed of 2 pi subunits. It exhibits a high activity for oxidation of long-chain aliphatic alcohols and aromatic alcohols and is less sensitive to pyrazole. This gene is localized to chromosome 4 in the cluster of alcohol dehydrogenase genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADH2; Alcohol dehydrogenase 2; Alcohol dehydrogenase 4; Alcohol dehydrogenase class II; Alcohol dehydrogenase class II pi chain; Alcohol dehydrogenase II; aldehyde reductase; All-trans-retinol dehydrogenase [NAD(+)] ADH4; class II alcohol dehydrogenase, pi subunit; class II type ADH; epididymis secretory protein Li 4
Gene Aliases: Ac1002; ADH-1; ADH-2; ADH2; ADH4; HEL-S-4
UniProt ID: (Human) P08319, (Mouse) Q9QYY9
Entrez Gene ID: (Human) 127, (Mouse) 26876, (Rat) 29646
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support