Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGTKGKVIKC KAAIAWEAGK PLCIEEVEVA PPKAHEVRIQ IIATSLCHTD ATVIDSKFEG LAFPVIVGHE AAGIVESIGP GVTNVKPGDK VIPLYAPLCR KCKFCLSPLT NLCGKISNLK SPASDQQLME DKTSRFTCKG KPVYHFFGTS TFSQYTVVSD INLAKIDDDA NLERVCLLGC GFSTGYGAAI NNAKVTPGST CAVFGLGGVG LSAVMGCKAA GASRIIGIDI NSEKFVKAKA LGATDCLNPR DLHKPIQEVI IELTKGGVDF ALDCAGGSET MKAALDCTTA GWGSCTFIGV AAGSKGLTVF PEELIIGRTI NGTFFGGWKS VDSIPKLVTD YKNKKFNLDA LVTHTLPFDK ISEAFDLMNQ GKSIRTILIF
This gene encodes class II alcohol dehydrogenase 4 pi subunit, which is a member of the alcohol dehydrogenase family. Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. Class II alcohol dehydrogenase is a homodimer composed of 2 pi subunits. It exhibits a high activity for oxidation of long-chain aliphatic alcohols and aromatic alcohols and is less sensitive to pyrazole. This gene is localized to chromosome 4 in the cluster of alcohol dehydrogenase genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alcohol dehydrogenase 2; Alcohol dehydrogenase 4; Alcohol dehydrogenase class II pi chain; aldehyde reductase; All-trans-retinol dehydrogenase [NAD(+)] ADH4; epididymis secretory protein Li 4
Gene Aliases: ADH-2; ADH2; ADH4; HEL-S-4
UniProt ID: (Human) P08319
Entrez Gene ID: (Human) 127
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support