Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSTTGQVIR CKAAILWKPG APFSIEEVEV APPKAKEVRI KVVATGLCGT EMKVLGSKHL DLLYPTILGH EGAGIVESIG EGVSTVKPGD KVITLFLPQC GECTSCLNSE GNFCIQFKQS KTQLMSDGTS RFTCKGKSIY HFGNTSTFCE YTVIKEISVA KIDAVAPLEK VCLISCGFST GFGAAINTAK VTPGSTCAVF GLGGVGLSVV MGCKAAGAAR IIGVDVNKEK FKKAQELGAT ECLNPQDLKK PIQEVLFDMT DAGIDFCFEA IGNLDVLAGR ADSTSLNWLL IIWQRS (1-295 aa encoded by B C039065)
This gene encodes class V alcohol dehydrogenase, which is a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. This gene is expressed in the stomach as well as in the liver, and it contains a glucocorticoid response element upstream of its 5' UTR, which is a steroid hormone receptor binding site. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alcohol dehydrogenase 6; alcohol dehydrogenase 6A (class V); aldehyde reductase; class V alcohol dehydrogenase
Gene Aliases: 1810009C16Rik; ADH-5; Adh5a; ADH6
UniProt ID: (Human) P28332, (Rat) Q5XI95
Entrez Gene ID: (Human) 130, (Rat) 310903, (Mouse) 69117, (Rat) 295498
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support