Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total ADI1.
This gene encodes an enzyme that belongs to the aci-reductone dioxygenase family of metal-binding enzymes, which are involved in methionine salvage. This enzyme may regulate mRNA processing in the nucleus, and may carry out different functions depending on its localization. A pseudogene that is located on chromosome 20 has been defined for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1,2-dihydroxy-3-keto-5-methylthiopentene dioxygenase; 1,2-dihydroxy-3-keto-5-methylthiopentene dioxygenase {ECO:0000255|HAMAP-Rule:MF_03154}; aci-reductone dioxygenase; aci-reductone dioxygenase (ARD)-like protein 1; Acireductone dioxygenase; Acireductone dioxygenase (Fe(2+)-requiring); acireductone dioxygenase (Fe(2+)-requiring) {ECO:0000255|HAMAP-Rule:MF_03154}; Acireductone dioxygenase (Ni(2+)-requiring); Androgen-responsive ARD-like protein 1; androgen-responsive gene encoding an ARD-like protein; ARD; ARD {ECO:0000255|HAMAP-Rule:MF_03154}; ARD'; fe-ARD; fe-ARD {ECO:0000255|HAMAP-Rule:MF_03154}; membrane-type 1 matrix metalloproteinase cytoplasmic tail binding protein-1; Membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1; membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1 {ECO:0000255|HAMAP-Rule:MF_03154}; MT1-MMP cytoplasmic tail-binding protein-1; MTCBP-1; MTCBP-1 {ECO:0000255|HAMAP-Rule:MF_03154}; Sip-L; submergence induced protein 2; Submergence-induced protein-like factor
Gene Aliases: ADI1; AL024210; Alp1; APL1; ARD; Fe-ARD; HMFT1638; MTCBP-1; MTCBP1; mtnD; Ni-ARD; SIPL
UniProt ID: (Human) Q9BV57, (Rat) Q562C9, (Mouse) Q99JT9
Entrez Gene ID: (Human) 55256, (Rat) 298934, (Mouse) 104923
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support