Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Catalyzes the reverse reaction of mono-ADP-ribosylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [Protein ADP-ribosylarginine] hydrolase; ADP-ribose-L-arginine cleaving enzyme; ADP-ribosylarginine hydrolase; ADP-ribosylhydrolase ARH1
Gene Aliases: ADPRH; ARH1
UniProt ID: (Human) P54922
Entrez Gene ID: (Human) 141
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support