Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MGQPGNGSAF LLAPNRSHAP DHDVTQQRDE VWVVGMGIVM SLIVLAIVFG NVLVITAIAK FERLQTVTNY FITSLACADL VMGLAVVPFG AAHILMKMWT FGNFWCEFWT SIDVLCVTAS IETLCVIAVD RYFAITSPFK YQSLLTKNKA RVIILMVWIV SGLTSFLPIQ MHWYRATHQE AINCYANETC CDFFTNQAYA IASSIVSFYV PLVIMVFVYS RVFQEAKRQL QKIDKSEGRF HVQNLSQVEQ DGRTGHGLRR SSKFCLKEHK ALKTLGIIMG TFTLCWLPFF IVNIVHVIQD NLIRKEVYIL LNWIGYVNSG FNPLIYCRSP DFRIAFQELL CLRRSSLKAY GNGYSSNGNT GEQSGYHVEQ EKENKLLCED LPGTEDFVGH QGTVPSDNID SQGRNCSTND SLL
Adrenergic receptors (ARs) are members of the 7-transmembrane domain G-protein-coupled receptor superfamily that bind the endogenous catecholamines epinephrine and norepinephrine. Pharmacological, structural, and molecular cloning data indicate significant heterogeneity within this receptor family. Nine receptor subtypes have been identified thus far including three alpha-1 AR subtypes (1A/D, 1B, and 1C), three alpha-2 ARs (2A, 2B, and 2C), and three beta AR subtypes (1, 2, and 3). ARs participate in either the onset or maintenance of several disease states including hypertension, cardiac dysfunction (congestive heart failure, ischemia, arrhythmias), diabetes, glaucoma, depression, and impotence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adrenergic, beta-2-, receptor, surface; adrenoceptor beta 2 surface; adrenoceptor beta 2, surface; AR beta2; Beta-2 adrenergic receptor; Beta-2 adrenoceptor; Beta-2 adrenoreceptor; beta2-Adrenoreceptor; catecholamine receptor; HGNC:286
Gene Aliases: ADRB2; ADRB2R; ADRBR; B2AR; BAR; BETA2AR
UniProt ID: (Human) P07550
Entrez Gene ID: (Human) 154
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support