Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAGGDHGSP DSYRSPLASR YASPEMCFVF SDRYKFRTWR QLWLWLAEAE QTLGLPITDE QIQEMKSNLE NIDFKMAAEE EKRLRHDVMA HVHTFGHCCP KAAGIIHLGA TSCYVGDNTD LIILRNALDL LLPKLARVIS RLADFAKERA SLPTLGFTHF QPAQLTTVGK RCCLWIQDLC MDLQNLKRVR DDLRFRGVKG TTGTQASFLQ LFEGDDHKVE QLDKMVTEKA GFKRAFIITG QTYTRKVDIE VLSVLASLGA SVHKICTDIR LLANLKEMEE PFEKQQIGSS AMPYKRNPMR SERCCSLARH LMTLVMDPLQ TASVQWFERT LDDSANRRIC LAEAFLTADT ILNTLQNISE GLVVYPKVIE RRIRQELPFM ATENIIMAMV KAGGSRQDCH EKIRVLSQQA ASVVKQEGGD NDLIERIQVD AYFSPIHSQL DHLLDPSSFT GRASQQVQRF LEEEVYPLLK PYESVMKVKA ELCL
ADSL (adenylosuccinate lyase), also known as AMPS, ASL or ASASE, is a 484 amino acid protein that is involved in both purine biosynthesis and in the formation of adenosine monophosphate (AMP) from inosine monophosphate. Expressed ubiquitously, ADSL catalyzes two key reactions in AMP biosynthesis, namely the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and the subsequent removal of fumarate from adenylosuccinate to yield AMP. Defects in the gene encoding ADSL are the cause of adenylosuccinase deficiency (ADSL deficiency), an autosomal recessive disorder characterized by epilepsy, growth retardation and muscular wasting. Multiple isoforms of ADSL exist due to alternative splicing events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylosuccinase; Adenylosuccinate lyase; ADSL; ASase
Gene Aliases: ADSL; AMPS; ASASE; ASL
UniProt ID: (Human) P30566
Entrez Gene ID: (Human) 158
Molecular Function:
lyase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support