Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody is predicted to react with bovine and porcine based on sequence homology.
ADSSL1 is a muscle isozyme of adenylosuccinate synthase (EC 6.3.4.4), which catalyzes the initial reaction in the conversion of inosine monophosphate (IMP) to adenosine monophosphate (AMP).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adenylosuccinate synthetase 1, muscle; Adenylosuccinate synthetase isozyme 1; Adenylosuccinate synthetase like 1; Adenylosuccinate synthetase, basic isozyme; Adenylosuccinate synthetase, muscle isozyme; Adenylosuccinate synthetase-like 1; adSS 1; AdSSL1; AMPSase 1; IMP--aspartate ligase 1; M-type adenylosuccinate synthetase
Gene Aliases: Adss; ADSS1; ADSSL1; AI528595
UniProt ID: (Human) Q8N142, (Mouse) P28650
Entrez Gene ID: (Human) 122622, (Mouse) 11565
Molecular Function: ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support