Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EQLTTASFPV GSKKNKGDSH LNVQVSNFKS GKGDSTLQVS SGLNENLTVN GGGWNEKSVK LSSQISAGEE KWNSVSPASA GKRKAEPSAW SQDTGDANTN GKDWGRSWSD RSIFSGIGST AEPVSQSTTS DYQWDVSRNQ PYIDDEWSGL NGLSSADPNS DWNAPAEEWG NWVDEERASL LKSQEPIPDD QKVSDDDKEK GEGALPTGKS KKKKKKKKKQ GEDNSTAQDT EELEKEIRED LPVNTSKTRP KQEKAFSLKT ISTSDPAEVL VKNSQPIKTL PPATSTEPSV ILSKSDSDKS SSQVPPILQE TDKSKSNTKQ NSVPPSQTKS ETSWESPKQI KKKKKARRET
Metadherin (Metastasis adhesion protein), also known as MTDH, LYsine-RIch CEACAM1 co-isolated (LYRIC), is a novel protein that localizes with the tight junction proteins ZO-1 and occludin in polarized epithelial cells. At the tight junction, it acts not as a structural component, but is rather recruited during the maturation of the tight junction complex. Metadherin is overexpressed in breast cancer tissue and breast tumor xenografts, while much lower levels are expressed in normal breast tissue. Metadherin binds to lung vasculature, one of the four common sites of breast cancer metastasis, through a C-terminal segment in the extracellular domain; blocking this lung-homing domain with antibodies or inhibiting metadherin with siRNA has been reported to inhibit breast cancer metastasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3D3/LYRIC; AEG-1; astrocyte elevated gene 1; Astrocyte elevated gene-1 protein; LYRIC/ MTDH; Lysine-rich CEACAM1 co-isolated protein; Metadherin; Metastasis adhesion protein; Protein LYRIC
Gene Aliases: 3D3; AEG-1; AEG1; LYRIC; LYRIC/3D3; MTDH
UniProt ID: (Human) Q86UE4
Entrez Gene ID: (Human) 92140
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support