Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: KNEPSFFPEQ KNRIIPPHQD NTHPSAPMPP PSVVILNSTL IHSNRKSKPE WSRDSHNPST VLASQASGQP NKMQTLTQDQ SQAKLEDFFV YPAEQPQIGE VEESNPSAKE D
AFF2 belongs to the AF4 family. Defects in AFF2 are the cause of FRAXE. The exact function of AFF2 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AF4/FMR2 family member 2; AF4/FMR2 family, member 2; FMR2P; fragile X E mental retardation syndrome protein; fragile X mental retardation 2 protein; Protein FMR-2; Protein Ox19
Gene Aliases: AFF2; FMR2; FMR2P; FRAXE; MRX2; OX19
UniProt ID: (Human) P51816
Entrez Gene ID: (Human) 2334
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support