Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SNYDGCCEG DVVQCIRDTS KVMNHICSKQ DSISSKIKEC CEKKIPERGQ CIINSNKDDR PKDLSLREGK FTDSENVCQE RDADPDTFFA KFTFEYSRRH PDLSIPELLR IVQIYKDLLR NCCNTENPPG CYRYAEDKFN ETTEKSLKMV QQECKHFQNL GKDGLKYHYL IRLTKIAPQL STEELVSLGE KMVTAFTTCC TLSEEFACVD NLADLVFGEL CGVNENRTIN PAVDHCCKTN FAFRRPCFES LKADKTYVPP PFSQDLFTFH ADMCQSQNEE LQRKTDRFLV NLVKLKHELT DEELQSLFTN FANVVDKCCK AESPEVCFNE ESPKIGN (264-599 aa encoded by BC109020 )
This gene is a member of the albumin gene family, which is comprised of four genes that localize to chromosome 4 in a tandem arrangement. These four genes encode structurally-related serum transport proteins that are known to be evolutionarily related. The protein encoded by this gene is regulated developmentally, expressed in the liver and secreted into the bloodstream.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Afamin; alpha albumin; Alpha-Alb; Alpha-albumin; MGC125338; MGC125339
Gene Aliases: AFM; ALB2; ALBA; ALF; alpha-Alb
UniProt ID: (Human) P43652, (Mouse) O89020
Entrez Gene ID: (Human) 173, (Mouse) 280662
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support