Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: SAKRKQEEK HLKMLRDMTG LPHNRKCFDC DQRGPTYVNM TVGSFVCTSC SGSLRGLNPP HRVKSISMTT FTQQEIEFLQ KHGNEVCKQI WLGLFDDRSS AIPDFRDPQK VKEFLQEKYE KKRWYVPPEQ AKVVASVHAS ISGSSASSTS STPEVKPLKS LLGDSAPTLH LNKGTPSQSP VVGRSQGQQQ EKKQFDLLSD LGSDIFAAPA PQSTATANFA NFAHFNSHAA QNSANADFAN FDAFGQSSGS SNFGGFPTAS HSPFQPQTTG GSAASVNANF AHFDNFPKSS SADFGTFNTS Q (4-303 aa encoded by BC030592)
HRB is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. HRB binds the activation domain of the human immunodeficiency virus Rev protein when Rev is assembled onto its RNA target, and is required for the nuclear export of Rev-directed RNAs. The protein encoded by this gene is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. This encoded protein binds the Rev activation domain when Rev is assembled onto its RNA target and can significantly enhance Rev activity when overexpressed. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arf-GAP domain and FG repeat-containing protein 1; arf-GAP domain and FG repeats-containing protein 1; HIV-1 Rev binding protein; HIV-1 Rev-binding protein; HIV-1 Rev-binding protein homolog; hRIP, Rev interacting protein; MGC116938; MGC116940; Nucleoporin-like protein RIP; Rab, Rev/Rex activation domain-binding protein; Rev-interacting protein; Rev/Rex activation domain-binding protein
Gene Aliases: AGFG1; Agfg1-ps1; AU045498; C130049H11Rik; C85612; D730048C23Rik; HRB; RAB; RIP
UniProt ID: (Human) P52594, (Mouse) Q8K2K6, (Rat) Q4KLH5
Entrez Gene ID: (Human) 3267, (Mouse) 15463, (Rat) 363266
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support