Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: KAAHFFSTLK EWPQTHQASI SYTGPTERPP NEPEETPVQR PSLYRRILRR LASYWAQPQD ALSQEVSPEV WKDVQLSTIE LSITTRNNQL DPTSKEDFLN ICIEPDTISK GDFITIGSRK VRNPKLHVEG TECLQASQCT LLIPEGAGGS FSIDSEEYEA MPVEVKLLPR KLQFFCDPRK REQMLTSPTQ; Positive Samples: 293T, SH-SY5Y, HepG2, HeLa, 22Rv1, BT-474, K-562, Rat brain; Cellular Location: Mitochondrion membrane
Lipid kinase that can phosphorylate both monoacylglycerol and diacylglycerol to form lysophosphatidic acid (LPA) and phosphatidic acid (PA), respectively. Does not phosphorylate sphingosine. Overexpression increases the formation and secretion of LPA, resulting in transactivation of EGFR and activation of the downstream MAPK signaling pathway, leading to increased cell growth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acylglycerol kinase, mitochondrial; FLJ10842; hAGK; HsMuLK; multi-substrate lipid kinase; Multiple substrate lipid kinase
Gene Aliases: AGK; CATC5; CTRCT38; MTDPS10; MULK; RGD1562046
UniProt ID: (Human) Q53H12
Entrez Gene ID: (Human) 55750, (Rat) 502749
Molecular Function: kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support