Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MTVFFKTLRN HWKKTTAGLC LLTWGGHWLY GKHCDNLLRR AACQEAQVFG NQLIPPNAQV KKATVFLNPA ACKGKARTLF EKNAAPILHL SGMDVTIVKT DYEGQAKKLL ELMENTDVII VAGGDGTLQE VVTGVLRRTD EATFSKIPIG FIPLGETSSL SHTLFAESGN KVQHITDATL AIVKGETVPL DVLQIKGEKE QPVFAMTGLR WGSFRDAGVK VSKYWYLGPL KIKAAHFFST LKEWPQTHQA SISYTGPTER PPNEPEETPV QRPSLYRRIL RRLASYWAQP QDALSQEVSP EVWKDVQLST IELSITTRNN QLDPTSKEDF LNICIEPDTI SKGDFITIGS RKVRNPKLHV EGTECLQASQ CTLLIPEGAG GSFSIDSEEY EAMPVEVKLL PRKLQFFCDP RKREQMLTSP TQ
Lipid kinase that can phosphorylate both monoacylglycerol and diacylglycerol to form lysophosphatidic acid (LPA) and phosphatidic acid (PA), respectively. Does not phosphorylate sphingosine. Overexpression increases the formation and secretion of LPA, resulting in transactivation of EGFR and activation of the downstream MAPK signaling pathway, leading to increased cell growth.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acylglycerol kinase, mitochondrial; FLJ10842; hAGK; HsMuLK; multi-substrate lipid kinase; Multiple substrate lipid kinase
Gene Aliases: AGK; CATC5; CTRCT38; MTDPS10; MULK
UniProt ID: (Human) Q53H12
Entrez Gene ID: (Human) 55750
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support