Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • AGL Antibodies

            Abnova

            AGL Polyclonal Antibody, MaxPab™

            View all (7) AGL antibodies
            Datasheet
            Protocols
            Questions & Answers
            Catalog # H00000178-B01P
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            465.00
            50 µg
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite AGL Polyclonal Antibody, MaxPab™

            • Antibody Testing Data (1)
            Application
            AGL Antibody in Western Blot (WB)
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            AGL Antibody (H00000178-B01P) in WB

            Western Blot analysis of AGL expression in transfected 293T cell line by AGL MaxPab polyclonal antibody. Lane 1: AGL transfected lysate(168.52 KDa). Lane 2: Non-transfected lysate. .
            View Product

            AGL Antibody in Western Blot (WB)

            Product Details

            H00000178-B01P

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:1,000
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Mouse / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            AGL (NP_000019.2, 1 a.a. approximately 1532 a.a) full-length human protein.
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Protein A

            Storage buffer

            PBS, pH 7.4

            Contains

            no preservative

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Product Specific Information

            Product may be used with Western Blot (Transfected lysate).

            The Isotype of this product is composed of an IgG Mixture.

            Immunogen sequence: MGHSKQIRIL LLNEMEKLEK TLFRLEQGYE LQFRLGPTLQ GKAVTVYTNY PFPGETFNRE KFRSLDWENP TEREDDSDKY CKLNLQQSGS FQYYFLQGNE KSGGGYIVVD PILRVGADNH VLPLDCVTLQ TFLAKCLGPF DEWESRLRVA KESGYNMIHF TPLQTLGLSR SCYSLANQLE LNPDFSRPNR KYTWNDVGQL VEKLKKEWNV ICITDVVYNH TAANSKWIQE HPECAYNLVN SPHLKPAWVL DRALWRFSCD VAEGKYKEKG IPALIENDHH MNSIRKIIWE DIFPKLKLWE FFQVDVNKAV EQFRRLLTQE NRRVTKSDPN QHLTIIQDPE YRRFGCTVDM NIALTTFIPH DKGPAAIEEC CNWFHKRMEE LNSEKHRLIN YHQEQAVNCL LGNVFYERLA GHGPKLGPVT RKHPLVTRYF TFPFEEIDFS MEESMIHLPN KACFLMAHNG WVMGDDPLRN FAEPGSEVYL RRELICWGDS VKLRYGNKPE DCPYLWAHMK KYTEITATYF QGVRLDNCHS TPLHVAEYML DAARNLQPNL YVVAELFTGS EDLDNVFVTR LGISSLIREA MSAYNSHEEG RLVYRYGGEP VGSFVQPCLR PLMPAIAHAL FMDITHDNEC PIVHRSAYDA LPSTTIVSMA CCASGSTRGY DELVPHQISV VSEERFYTKW NPEALPSNTG EVNFQSGIIA ARCAISKLHQ ELGAKGFIQV YVDQVDEDIV AVTRHSPSIH QSVVAVSRTA FRNPKTSFYS KEVPQMCIPG KIEEVVLEAR TIERNTKPYR KDENSINGTP DITVEIREHI QLNESKIVKQ AGVATKGPNE YIQEIEFENL SPGSVIIFRV SLDPHAQVAV GILRNHLTQF SPHFKSGSLA VDNADPILKI PFASLASRLT LAELNQILYR CESEEKEDGG GCYDIPNWSA LKYAGLQGLM SVLAEIRPKN DLGHPFCNNL RSGDWMIDYV SNRLISRSGT IAEVGKWLQA MFFYLKQIPR YLIPCYFDAI LIGAYTTLLD TAWKQMSSFV QNGSTFVKHL SLGSVQLCGV GKFPSLPILS PALMDVPYRL NEITKEKEQC CVSLAAGLPH FSSGIFRCWG RDTFIALRGI LLITGRYVEA RNIILAFAGT LRHGLIPNLL GEGIYARYNC RDAVWWWLQC IQDYCKMVPN GLDILKCPVS RMYPTDDSAP LPAGTLDQPL FEVIQEAMQK HMQGIQFRER NAGPQIDRNM KDEGFNITAG VDEETGFVYG GNRFNCGTWM DKMGESDRAR NRGIPATPRD GSAVEIVGLS KSAVRWLLEL SKKNIFPYHE VTVKRHGKAI KVSYDEWNRK IQDNFEKLFH VSEDPSDLNE KHPNLVHKRG IYKDSYGASS PWCDYQLRPN FTIAMVVAPE LFTTEKAWKA LEIAEKKLLG PLGMKTLDPD DMVYCGIYDN ALDNDNYNLA KGFNYHQGPE WLWPIGYFLR AKLYFSRLMG PETTAKTIVL VKNVLSRHYV HLERSPWKGL PELTNENAQY CPFSCETQAW SIATILETLY DL

            Target Information

            This gene encodes the glycogen debrancher enzyme which is involved in glycogen degradation. This enzyme has two independent catalytic activities which occur at different sites on the protein: a 4-alpha-glucotransferase activity and a amylo-1,6-glucosidase activity. Mutations in this gene are associated with glycogen storage disease although a wide range of enzymatic and clinical variability occurs which may be due to tissue-specific alternative splicing. Alternatively spliced transcripts encoding different isoforms have been described.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: 1-Apr; 4-glucantransferase; amylo-1, 6-glucosidase, 4-alpha-glucanotransferase; amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferaseprovided; Glycogen debrancher; Glycogen debranching enzyme; glycogen debranching protein; Oligo-1

            View more View less

            Gene Aliases: AGL; GDE

            View more View less

            UniProt ID: (Human) P35573

            View more View less

            Entrez Gene ID: (Human) 178

            View more View less

            Function(s)

            glycogen debranching enzyme activity 4-alpha-glucanotransferase activity amylo-alpha-1,6-glucosidase activity protein binding polysaccharide binding polyubiquitin binding

            Process(es)

            glycogen biosynthetic process glycogen catabolic process response to nutrient response to glucocorticoid
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-7vhdw:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0