Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is an acyltransferase that converts lysophosphatidic acid into phosphatidic acid, which is the second step in the de novo phospholipid biosynthetic pathway. The encoded protein may be an integral membrane protein. Two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-acyl-sn-glycerol-3-phosphate acyltransferase gamma; 1-acylglycerol-3-phosphate O-acyltransferase 3; 1-AGP acyltransferase 3; 1-AGPAT 3; LPAAT-gamma; Lysophosphatidic acid acyltransferase gamma; lysophosphatidic acid acyltransferase-gamma1
Gene Aliases: AGPAT3; LPAAT-GAMMA1; LPAAT3; UNQ759/PRO1490
UniProt ID: (Human) Q9NRZ7
Entrez Gene ID: (Human) 56894
Molecular Function:
acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support