Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDLAGLLKSQ FLCHLVFCYV FIASGLIINT IQLFTLLLWP INKQLFRKIN CRLSYCISSQ LVMLLEWWSG TECTIFTDPR AYLKYGKENA IVVLNHKFEI DFLCGWSLSE RFGLLGGSKV LAKKELAYVP IIGWMWYFTE MVFCSRKWEQ DRKTVATSLQ HLRDYPEKYF FLIHCEGTRF TEKKHEISMQ VARAKGLPRL KHHLLPRTKG FAITVRSLRN VVSAVYDCTL NFRNNENPTL LGVLNGKKYH ADLYVRRIPL EDIPEDDDEC SAWLHKLYQE KDAFQEEYYR TGTFPETPMV PPRRPWTLVN WLFWASLVLY PFFQFLVSMI RSGSSLTLAS FILVFFVASV GVRWMIGVTE IDKGSAYGNS DSKQKLND
This gene encodes a member of the 1-acylglycerol-3-phosphate O-acyltransferase family. This integral membrane protein converts lysophosphatidic acid to phosphatidic acid, the second step in de novo phospholipid biosynthesis. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-acyl-sn-glycerol-3-phosphate acyltransferase delta; 1-acylglycerol-3-phosphate O-acyltransferase 4; 1-acylglycerol-3-phosphate O-acyltransferase 4 (lysophosphatidic acid acyltransferase, delta); 1-AGP acyltransferase 4; 1-AGPAT 4; LPAAT-delta; Lysophosphatidic acid acyltransferase delta; lysophosphatidic acid acyltransferase-delta (LPAAT-delta)
Gene Aliases: 1-AGPAT4; AGPAT4; dJ473J16.2; LPAAT-delta; UNQ499/PRO1016
UniProt ID: (Human) Q9NRZ5
Entrez Gene ID: (Human) 56895
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support