Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SFGIRKLYMK SLLKIFAWAT LRMERGAKEK NHQLYKPYTN GIIAKDPTSL EEEIKEIRRS GSSKALDNTP EFELSDIFYF CRKGMETIMD DEVTKRFSA
Lysophosphatidic acid acyltransferases (EC 2.3.1.51) catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA). LPA and PA are involved in signal transduction and lipid biosynthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-acyl-sn-glycerol-3-phosphate acyltransferase zeta; 1-acylglycerol-3-phosphate O-acyltransferase 6; 1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acid acyltransferase, zeta); 1-AGP; 1-AGP acyltransferase 6; Acyl-CoA:glycerol-3-phosphate acyltransferase 4; Glycerol-3-phosphate acyltransferase 4; glycerol-3-phosphate acyltransferase 6; glycerol-3-phosphate acyltransferase-4; LPAAT-zeta; Lysophosphatidic acid acyltransferase zeta; Testis spermatogenesis apoptosis-related protein 7; TSARG7
Gene Aliases: 1-AGPAT 6; AGPAT6; GPAT4; LPAAT-zeta; LPAATZ; TSARG7; UNQ551/PRO1108
UniProt ID: (Human) Q86UL3
Entrez Gene ID: (Human) 137964
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support