Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Sequence of this protein is as follows: KARRAASAA
This gene is a member of the FAD-binding oxidoreductase/transferase type 4 family. It encodes a protein that catalyzes the second step of ether lipid biosynthesis in which acyl-dihydroxyacetonephosphate (DHAP) is converted to alkyl-DHAP by the addition of a long chain alcohol and the removal of a long-chain acid anion. The protein is localized to the inner aspect of the peroxisomal membrane and requires FAD as a cofactor. Mutations in this gene have been associated with rhizomelic chondrodysplasia punctata, type 3 and Zellweger syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aging-associated gene 5 protein; aging-associated protein 5; Alkyl-DHAP synthase; alkyl-dihydroxyacetonephosphate synthase; alkyldihydroxyacetone phosphate synthase; Alkyldihydroxyacetonephosphate synthase, peroxisomal; alkylglycerone phosphate synthase-like; Alkylglycerone-phosphate synthase
Gene Aliases: 5832437L22; 9930035G10Rik; AAG5; ADAP-S; Adaps; ADAS; ADHAPS; ADPS; Ads; AGPS; ALDHPSY; AW123847; bs2; RCDP3
UniProt ID: (Human) O00116, (Rat) Q9EQR2, (Mouse) Q8C0I1
Entrez Gene ID: (Human) 8540, (Rat) 84114, (Mouse) 228061
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support