Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: PTAVFGTLAS LYLGALDHTA DRLQAILGVP WKDKNCTSRL DAHKVLSALQ AVQGLLVAQG RADSQAQLLL STVVGVFTAP GLHLKQPFVQ GLALYTPVVL PRSLDFTELD VAAEKIDRFM QAVTGWKTGC SLTGASVDST LAFNTYVHFQ GKMKGFSLLA EPQEFWVDNS TSVSVPMLSG MGTFQHWSDI QDNFSVTQVS FTESACLLLI QPHYASDLDK VEGLTFQQNS LNWMKKLSPR TIHLTMPQLV LQGSYDLQDL LAQAELPAIL HTELNLQKLS NDRIRVGEVL NSIFFELEAD EREPTESTQQ LNKPEVLEVT LNRPFLFAVY DQSATALHFL GRVANPLSTA
The protein encoded by AGT, pre-angiotensinogen or angiotensinogen precursor, is expressed in the liver and is cleaved by the enzyme renin in response to lowered blood pressure. The resulting product, angiotensin I, is then cleaved by angiotensin converting enzyme (ACE) to generate the physiologically active enzyme angiotensin II. The protein is involved in maintaining blood pressure and in the pathogenesis of essential hypertension and preeclampsia. Mutations in this gene are associated with susceptibility to essential hypertension, and can cause renal tubular dysgenesis, a severe disorder of renal tubular development. Defects in this gene have also been associated with non-familial structural atrial fibrillation, and inflammatory bowel disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-1 antiproteinase, antitrypsin; angiotensin I; angiotensin II; Angiotensinogen; angiotensinogen (serpin peptidase inhibitor, clade A, member 8); pre-angiotensinogen; serine (or cysteine) proteinase inhibitor; Serpin A8; serpin peptidase inhibitor, clade A, member 8
Gene Aliases: AGT; ANHU; SERPINA8
UniProt ID: (Human) P01019
Entrez Gene ID: (Human) 183, (Pig) 100157073
Molecular Function:
protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support