Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This gene is expressed only in the liver and the encoded protein is localized mostly in the peroxisomes, where it is involved in glyoxylate detoxification. Mutations in this gene, some of which alter subcellular targetting, have been associated with type I primary hyperoxaluria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AGT; Alanine--glyoxylate aminotransferase; alanine-glyoxylate aminotransferase 1; angiotensin receptor 2; hepatic peroxisomal alanine:glyoxylate aminotransferase; L-alanine: glyoxylate aminotransferase 1; Serine--pyruvate aminotransferase; Serine--pyruvate aminotransferase, mitochondrial; serine--pyruvate aminotransferase, peroxisomal; serine-pyruvate aminotransferase; serine:pyruvate aminotransferase; serine:pyruvate aminotransferase SPT; serine:pyruvate/alanine:glyoxylate aminotransferase; SPT
Gene Aliases: AGT; AGT1; AGXT; AGXT1; PH1; SPAT; SPT; TLH6
UniProt ID: (Human) P21549, (Rat) P09139, (Mouse) O35423
Entrez Gene ID: (Human) 189, (Rat) 24792, (Mouse) 11611
Molecular Function: transaminase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support