Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MASHKLLVTP PKALLKPLSI PNQLLLGPGP SNLPPRIMAA GGLQMIGSMS KDMYQIMDEI KEGIQYVFQT RNPLTLVISG SGHCALEAAL VNVLEPGDSF LVGANGIWGQ RAVDIGERIG ARVHPMTKDP GGHYTLQEVE EGLAQHKPVL LFLTHGESST GVLQPLDGFG ELCHRYKCLL LVDSVASLGG TPLYMDRQGI DILYSGSQKA LNAPPGTSLI SFSDKAKKKM YSRKTKPFSF YLDIKWLANF WGCDDQPRMY HHTIPVISLY SLRESLALIA EQGLENSWRQ HREAAAYLHG RLQALGLQLF VKDPALRLPT VTTVAVPAGY DWRDIVSYVI DHFDIEIMGG LGPSTGKVLR IGLLGCNATR ENVDRVTEAL RAALQHCPKK KL
This gene is expressed only in the liver and the encoded protein is localized mostly in the peroxisomes, where it is involved in glyoxylate detoxification. Mutations in this gene, some of which alter subcellular targetting, have been associated with type I primary hyperoxaluria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AGT; Alanine--glyoxylate aminotransferase; hepatic peroxisomal alanine:glyoxylate aminotransferase; L-alanine: glyoxylate aminotransferase 1; Serine--pyruvate aminotransferase; serine-pyruvate aminotransferase; serine:pyruvate aminotransferase; SPT
Gene Aliases: AGT; AGT1; AGXT; AGXT1; PH1; SPAT; SPT; TLH6
UniProt ID: (Human) P21549
Entrez Gene ID: (Human) 189
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support