Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MSMPDAMPLP GVGEELKQAK EIEDAEKYSF MATVTKAPKK QIQFADDMQE FTKFPTKTGR RSLSRSISQS STDSYSSAAS YTDSSDDEVS PREKQQTNSK G
The protein encoded by this gene interacts with inositol 1,4,5-trisphosphate receptor, type 1 and may be involved in the conversion of S-adenosyl-L-homocysteine to L-homocysteine and adenosine. Several transcript variants encoding two different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adenosylhomocysteinase-like 1; AdoHcyase 2; DC-expressed AHCY-like molecule; dendritic cell expressed AHCY-like protein; inositol 1,4,5-trisphosphate receptor-binding protein; IP(3)Rs binding protein released with IP(3); IRBIT; protein phosphatase 1, regulatory subunit 78; Putative adenosylhomocysteinase 2; s adenosylhomocysteine hydrolase; S-adenosyl homocysteine hydrolase homolog; S-adenosyl-L-homocysteine hydrolase 2; S-adenosylhomocysteine hydrolase-like protein 1
Gene Aliases: AHCYL1; DCAL; IRBIT; PPP1R78; PRO0233; XPVKONA
UniProt ID: (Human) O43865
Entrez Gene ID: (Human) 10768
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support