Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPTAESEAKV KTKVRFEELL KTHSDLMREK KKLKKKLVRS EENISPDTIR SNLHYMKETT SDDPDTIRSN LPHIKETTSD DVSAANTNNL KKSTRVTKNK LRNTQLATEN PNGDASVEED KQGKPNKKVI KTVPQLTTQD LKPETPENKV DSTHQKTHTK PQPGVDHQKS EKANEGREET DLEEDEELMQ AYQCHVTEEM AKEIKRKIRK KLKEQLTYFP SDTLFHDDKL SSEKRKKKKE VPVFSKAETS TLTISGDTVE GEQKKESSVR SVSSDSHQDD EISSMEQSTE DSMQDDTKPK PKKTKKKTKA VADNNEDVDG DGVHEITSRD SPVYPKCLLD DDLVLGVYIH RTDRLKSDFM ISHPMVKIHV VDEHTGQYVK KDDSGRPVSS YYEKENVDYI LPIMTQPYDF KQLKSRLPEW EEQIVFNENF PYLLRGSDES PKVILFFEIL DFLSVDEIKN NSEVQNQECG FRKIAWAFLK LLGANGNANI NSKLRLQLYY PPTKPRSPLS VVEAFEWWSK CPRNHYPSTL YVTVRGLKVP DCIKPSYRSM MALQEEKGKP VHCERHHESS SVDTEPGLEE SKEVIKWKRL PGQENEDVFV LISPTMEEY
This gene is apparently required for both cerebellar and cortical development in humans. This gene mutations cause specific forms of Joubert syndrome-related disorders. Joubert syndrome (JS) is a recessively inherited developmental brain disorder with several identified causative chromosomal loci. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: abelson helper integration site 1 protein; Abelson helper integration site 1 protein homolog; AHI-1; contatins SH3 and WD40 domains; Jouberin; RP1-32B1.2
Gene Aliases: 1700015F03Rik; AHI-1; AHI1; D10Bwg0629e; dJ71N10.1; JBTS3; ORF1
UniProt ID: (Human) Q4FD35
Entrez Gene ID: (Human) 54806, (Mouse) 52906
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support