Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MEKEEETTR ELLLPNWQGS GSHGLTIAQR DDGVFVQEVT QNSPAARTGV VKEGDQIVGA TIYFDNLQSG EVTQLLNTMG HHTVGLKLHR KGDRSPEPGQ TWTREVFSSC SSEVVLNTPQ PSALECKDQN KQKEASSQAG AVSVSTPNAG (1-149 aa encoded by BC000926)
AHNAK1 (Desmoyokin) is a large (700 kDa) scaffold protein that translocates to the plasma membrane after an increas of extracellular calcium level or upon proteinkinase C activation and regulates extracellular calcium influx mediated by L-type Ca2+ channels. AHNAK1 has been implicated in diverse signal transduction proceses affecting cell differentiation and proliferation. In response to calcium-dependent intercellular contacts AHNAK1 forms multimeric complexes in the plasma membrane, connected with actin and annexin 2/S100A10 assemblies and is thus involved in organization of the plasma membrane architecture. In epithelial cells, AHNAK1 is localized in cytoplasm or is membrane-associated, but in cells of nonepithelial origin AHNAK1 is predominantly nuclear; it has a weak DNA-binding activity and associates with the DNA-ligase IV-XRCC4 complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AHNAK-related; Desmoyokin; MGC5395; Neuroblast differentiation-associated protein AHNAK
Gene Aliases: AHNAK; AHNAKRS; PM227
UniProt ID: (Human) Q09666
Entrez Gene ID: (Human) 79026
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support