Save $650 & get a lab-themed hidden object puzzle

Save and claim

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • AMID Antibodies

            Proteintech

            AIFM2 Polyclonal Antibody

            68 References
            View all (18) AMID antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 20886-1-AP
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            492.00
            150 µL
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite AIFM2 Polyclonal Antibody

            • Antibody Testing Data (6)
            Application
            AIFM2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 6

            AIFM2 Antibody (20886-1-AP) in IHC (P)

            Immunohistochemical analysis of paraffin-embedded human liver tissue slide using (Product # 20886-1-AP) (AIFM2/ FSP1 antibody) at dilution of 1:200 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0).
            View Product

            AIFM2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            AIFM2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            AIFM2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            AIFM2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            AIFM2 Antibody in Western Blot (WB)
            AIFM2 Antibody in Immunoprecipitation (IP)

            Product Details

            20886-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:1,500
            View 48 publications 48 publications

            Immunohistochemistry (IHC)

            -
            View 11 publications 11 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:50-1:500
            -

            Immunocytochemistry (ICC/IF)

            -
            View 8 publications 8 publications

            Immunoprecipitation (IP)

            0.5-4.0 µg
            -

            Not applicable (N/A)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human, Mouse

            Published species

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            AIFM2 Fusion Protein Ag13412 (1-373 aa encoded by BC023601)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.27 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: MGSQVSVES GALHVVIVGG GFGGIAAASQ LQALNVPFML VDMKDSFHHN VAALRASVET GFAKKTFISY SVTFKDNFRQ GLVVGIDLKN QMVLLQGGEA LPFSHLILAT GSTGPFPGKF NEVSSQQAAI QAYEDMVRQV QRSRFIVVVG GGSAGVEMAA EIKTEYPEKE VTLIHSQVAL ADKELLPSVR QEVKEILLRK GVQLLLSERV SNLEELPLNE YREYIKVQTD KGTEVATNLV ILCTGIKINS SAYRKAFESR LASSGALRVN EHLQVEGHSN VYAIGDCADV RTPKMAYLAG LHANIAVANI VNSVKQRPLQ AYKPGALTFL LSMGRNDGVG QISGFYVGRL MVRLTKSRDL FVSTSWKTMR QSPP (1-373 aa encoded by BC023601)

            Target Information

            The p53 tumor suppressor protein induces cell cycle arrest or apoptosis in response to cellular stresses. PRG3 (p53-responsive gene 3), is induced specifically under p53-dependent apoptotic conditions in human colon cancer cells. PRG3 has significant homology to bacterial oxidoreductases and the apoptosis-inducing factor, AIF. Unlike AIF, PRG3 localizes in the cytoplasm and its ectopic expression induces apoptosis. The PRG3 gene is thus a novel p53 target gene in a p53-dependent apoptosis pathway.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: AMID; apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer of death; apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death; apoptosis-inducing factor 2; Apoptosis-inducing factor homologous mitochondrion-associated inducer of death; apoptosis-inducing factor, mitochondrion-associated, 2; apoptosis-inducing factor-like mitchondrion-associated inducer of death; apoptosis-inducing factor-like mitochondrion-associated inducer of death; Ferroptosis suppressor protein 1; FSP1; MBP2; MBPH; MGC126662; MGC141971; p53-responsive gene 3 protein; RP11-367H5.2

            View more View less

            Gene Aliases: 5430437E11Rik; AIFM2; AMID; D730001I10Rik; PRG3

            View more View less

            UniProt ID: (Human) Q9BRQ8, (Mouse) Q8BUE4

            View more View less

            Entrez Gene ID: (Human) 84883, (Mouse) 71361

            View more View less

            Function(s)

            DNA binding electron-transferring-flavoprotein dehydrogenase activity flavin adenine dinucleotide binding NADH dehydrogenase (ubiquinone) activity N-ethylmaleimide reductase activity oxidoreductase activity reduced coenzyme F420 dehydrogenase activity sulfur oxygenase reductase activity malolactic enzyme activity NADPH:sulfur oxidoreductase activity epoxyqueuosine reductase activity oxidoreductase metabolite interconversion enzyme

            Process(es)

            apoptotic mitochondrial changes regulation of apoptotic process positive regulation of apoptotic process oxidation-reduction process
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-nfgkx:80/100.66.77.49:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0