Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AHSVSVVEL EETPFRRFLG ERVGRALMKM FENNRVKFYM QTEVSELRGQ EGKLKEVVLK SSKVVRADVC VVGIGAVPAT GFLRQSGIGL DSRGFIPVNK MMQTNVPGVF AAGDAVTFPL AWRNNRKVNI PHWQMAHAQG RVAAQNMLAQ EAEMSTVPYL WTAMFGKSLR YAGYGEGFDD VIIQGDLEEL KFVAFYTKGD EVIAVASMNY DPIVSKVAEV LASGRAIRKR EVETGDMSWL TGKGS (355-598 aa encoded by BC032485)
Apoptosis, also known as programmed cell death, plays major roles in development and normal tissue turnover in addition to tumor formation. Recently a protein similar to the apoptosis-inducing factor (AIF) was cloned and designated AIFL (also known as AIFM3). AIFM3 is expressed ubiquitously and is predominantly localized to the inner membranes of mitochondria. Unlike AIF, AIFM3 does not translocate to the nucleus upon induction of apoptosis. However, overexpression of AIFM3, like AIF, induced cytochrome c release from the mitochondria, cleavage of caspase 3, and ultimately apoptosis, indicating AIFM3 induces apoptosis through caspase activation. Multiple isoforms of AIFM3 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apoptosis-inducing factor 3; apoptosis-inducing factor like; apoptosis-inducing factor, mitochondrion-associated, 3; Apoptosis-inducing factor-like protein
Gene Aliases: 2810401C16Rik; AI840249; AIFL; AIFM3; RGD1306028
UniProt ID: (Human) Q96NN9, (Mouse) Q3TY86
Entrez Gene ID: (Human) 150209, (Mouse) 72168, (Rat) 303786
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support