Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 1
Immunogen sequence: LGCGAASGTP SGILYEPPAE KEQKVLIQSV SCTGTEDTLA QCEQEEVYDC
CD5L, also known as CD5 antigen-like, is a protein that is expressed on the surface of various immune cells, including T cells, B cells, and natural killer cells. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including autoimmune disorders, cancer, and infectious diseases. CD5L is known to modulate T cell activation and cytokine production, and has been shown to have anti-inflammatory effects.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: apoptosis inhibitor 6; Apoptosis inhibitor expressed by macrophages; apoptosis inhibitor of macrophage; APOPTOSIS INHIBITOR OF MACROPHAGES; CD5 antigen like (scavenger receptor cysteine rich family); CD5 antigen-like; CD5 antigen-like (scavenger receptor cysteine rich family); CD5 molecule-like; CT-2; hAIM; Highly similar to ANTIGEN WC1.1 [Bos taurus]; IgM-associated peptide; SCAVENGER RECEPTOR CYSTEINE-RICH FAMILY; SP-alpha
Gene Aliases: AIM; API6; CD5L; CT-2; hAIM; PRO229; SP-ALPHA; Spalpha; UNQ203/PRO229
UniProt ID: (Human) O43866
Entrez Gene ID: (Human) 922
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support