Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MESKYKEILL LTGLDNITDE ELDRFKFFLS DEFNIATGKL HTANRIQVAT LMIQNAGAVS AVMKTIRIFQ KLNYMLLAKR LQEEKEKVDK QYKSVTKPKP LSQAEMSPAA SAAIRNDVAK QRAAPKVSPH VKPEQKQMVA QQESIREGFQ KRCLPVMVLK AKKPFTFETQ EGKQEMFHAT VATEKEFFFV KVFNTLLKDK FIPKRIIIIA RYYRHSGFLE VNSASRVLDA ESDQKVNVPL NVIRKAGETP KINTLQTQPL GTIVNGLFVV QKVTEKKKNI LFDLSDNTGK MEVLGVRNED TMKCKEGDKV RLTFFTLSKN GEKLQLTSGV HSTIKVIKAK KKKHREVKRT NSSQLV
AIM2 is a member of the IFI20X /IFI16 family. It plays a putative role in tumorigenic reversion and may control cell proliferation. Interferon-gamma induces expression of AIM2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Absent in melanoma 2; Interferon-inducible protein AIM2; RP11-520H16.3
Gene Aliases: AIM2; PYHIN4
UniProt ID: (Human) O14862
Entrez Gene ID: (Human) 9447
Molecular Function: DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support