Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
Human ARA9 shares 95% amino acid (aa) sequence identity with both mouse and rat ARA9.
Positive Control - WB: human HELA whole cell, human HEPG2 whole cell, human placenta tissue, human CACO-2 whole cell, human U20S whole cell, monkey COS-7 whole cell, human K562 whole cell, rat heart tissue, rat skeletal muscle tissue, rat brain tissue, rat spleen tissue, mouse brain tissue, mouse HEPA1-6 whole cell. IHC: human intestinal cancer tissue, human tonsil tissue, rat spleen tissue. ICC/IF: A431 cell. Flow: A549 cell.|Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AH receptor-interacting protein; AIP; Aryl-hydrocarbon receptor-interacting protein; HBV X-associated protein 2; HBV-X associated protein; Immunophilin homolog ARA9; Immunophilin XAP2; XAP-2
Gene Aliases: AA408703; AIP; ARA9; AW476050; D19Bwg1412e; FKBP16; FKBP37; SMTPHN; XAP-2; XAP2
UniProt ID: (Human) O00170, (Mouse) O08915, (Rat) Q5FWY5
Entrez Gene ID: (Human) 9049, (Mouse) 11632, (Rat) 282827
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support