Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEEKLKKTN IIFVVGGPGS GKGTQCEKIV QKYGYTHLST GDLLRSEVSS GSARGKKLSE IMEKGQLVPL ETVLDMLRDA MVAKVNTSKG FLIDGYPREV QQGEEFERRI GQPTLLLYVD AGPETMTQRL LKRGETSGRV DDNEETIKKR LETYYKATEP VIAFYEKRGI VRKVNAEGSV DSVFSQVCTH LDALK (1-194 aa encoded by BC001116)
Adenylate kinase is an enzyme involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate group among adinine nucleotides. Three isozymes of adenylate kinase have been identified in vertebrates, adenylate isozyme 1 (AK1), 2 (AK2) and 3 (AK3). AK1 is found in the cytosol of skeletal muscle, brain and erythrocytes, whereas AK2 and AK3 are found in the mitochondria of other tissues including liver and heart. AK1 was identified because of its association with a rare genetic disorder causing nonspherocytic hemolytic anemia where a mutation in the AK1 gene was found to reduce the catalytic activity of the enzyme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylate kinase isoenzyme 1; adenylate kinase isoenzyme 1 {ECO:0000255|HAMAP-Rule:MF_03171}; Adenylate monophosphate kinase; adenylate monophosphate kinase {ECO:0000255|HAMAP-Rule:MF_03171}; AK 1; AK 1 {ECO:0000255|HAMAP-Rule:MF_03171}; ATP-AMP transphosphorylase 1; ATP-AMP transphosphorylase 1 {ECO:0000255|HAMAP-Rule:MF_03171}; ATP:AMP phosphotransferase; ATP:AMP phosphotransferase {ECO:0000255|HAMAP-Rule:MF_03171}; cytosolic adenylate kinase; Myokinase; myokinase {ECO:0000255|HAMAP-Rule:MF_03171}; testis secretory sperm binding protein Li 58j
Gene Aliases: Ak 1; Ak-1; AK1; B430205N08Rik; HTL-S-58j
UniProt ID: (Human) P00568, (Mouse) Q9R0Y5, (Rat) P39069
Entrez Gene ID: (Human) 203, (Mouse) 11636, (Rat) 24183
Molecular Function:
nucleotide kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support