Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 27
Immunogen sequence: MGASARLLR AVIMGAPGSG KGTVSSRITT HFELKHLSSG DLLRDNMLRG TEIGVLAKAF IDQGKLIPDD VMTRLALHEL KNLTQYSWLL DGFPRTLPQA EALDRAYQID TVINLNVPFE VIKQRLTARW IHPASGRVYN IEFNPPKTVG IDDLTGEPLI QREDDKPETV IKRLKAYEDQ TKPVLEYYQK KGVLETFSGT ETNKIWPYVY AFLQTKVPQR SQKASVTP (1-227 aa encoded by BC013771)
This gene encodes a member of the adenylate kinase family of enzymes. The encoded protein is localized to the mitochondrial matrix. Adenylate kinases regulate the adenine and guanine nucleotide compositions within a cell by catalyzing the reversible transfer of phosphate group among these nucleotides. Five isozymes of adenylate kinase have been identified in vertebrates. Expression of these isozymes is tissue-specific and developmentally regulated. A pseudogene for this gene has been located on chromosome 17. Three transcript variants encoding the same protein have been identified for this gene. Sequence alignment suggests that the gene defined by NM_013410, NM_203464, and NM_001005353 is located on chromosome 1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylate kinase 3; Adenylate kinase 3 alpha-like 1; adenylate kinase 3 alpha-like 1 {ECO:0000255|HAMAP-Rule:MF_03169}; adenylate kinase 3 {ECO:0000255|HAMAP-Rule:MF_03169}; adenylate kinase 6, adenylate kinase 3 like 1; Adenylate kinase isozyme 3; AK 3; AK 3 {ECO:0000255|HAMAP-Rule:MF_03169}; AK3; AK4; GTP:AMP phosphotransferase AK3, mitochondrial; GTP:AMP phosphotransferase AK3, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03169}; GTP:AMP phosphotransferase mitochondrial; GTP:AMP phosphotransferase, mitochondrial
Gene Aliases: AA407498; AI506714; AK-3; AK3; Ak3l; AK3L1; AK6; AKL3L; AKL3L1; FIX
UniProt ID: (Human) Q9UIJ7, (Rat) P29411, (Mouse) Q9WTP7
Entrez Gene ID: (Human) 50808, (Rat) 26956, (Mouse) 56248
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support