Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAEEEETAAL TEKVIRTQRV FINLLDSYSS GNIGKFLSNC VVGASLEEIT EEEEEEDENK SAMLEASSTK VKEGTFQIVG TLSKPDSPRP DFAVETYSAI SREDLLMRLL ECDVIIYNIT ESSQQMEEAI WAVSALSEEV SHFEKRKLFI LLSTVMTWAR SKALDPEDSE VPFTEEDYRR RKSHPNFLDH INAEKMVLKF GKKARKFAAY VVAAGLQYGA EGGMLHTFFK MAWLGEIPAL PVFGDGTNVI PTIHVLDLAG VIQNVIDHVP KPHYLVAVDE SVHTLEDIVK CISKNTGPGK IQKIPRENAY LTKDLTQDCL DHLLVNLRME ALFVKENFNI RWAAQTGFVE NINTILKEYK QSRGLMPIKI CILGPPAVGK SSIAKELAKY YKLHHIQLKD VISEAIAKLE AIVAPNDVGE GEEEVEEEEE EENVEDAQEL LDGIKESMEQ NAGQLDDQYI IRFMKEKLKS MPCRNQGYIL DGFPKTYDQA KDLFNQEDEE EEDDVRGRMF PFDKLIIPEF VCALDASDEF LKERVINLPE SIVAGTHYSQ DRFLRALSNY RDINIDDETV FNYFDELEIH PIHIDVGKLE DAQNRLAIKQ LIKETGEPRN YGLTDEEKAE EERKAAEERL AREAAEEAER EHQEAVEMAE KIARWE
The adenylate kinases (AK) are a family of structurally and functionally related enzymes that catalyze a similar reaction, MgNTP + AMP = MgNDP + ADP (N = A or G). The AK enzymes are important for maintenance of homeostasis of the adenine and guanine nucleotide pools. AK1 is a cytosolic enzyme for which ATP is the substrate. AK2 catalyzes the same reaction as AK1, but it is localized in the mitochondrial intermembrane space. AK3 is present in the mitochondrial matrix and prefers GTP over ATP as the substrate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylate kinase 7; AK 7; ATP-AMP transphosphorylase 7; putative adenylate kinase 7
Gene Aliases: AK 7; AK7
UniProt ID: (Human) Q96M32
Entrez Gene ID: (Human) 122481
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support