Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDQGYGGYGA WSAGPANTQG AYGTGVASWQ GYENYNYYGA QNTSVTTGAT YSYGPASWEA AKANDGGLAA GAPAMHMASY GPEPCTDNSD SLIAKINQRL DMMSKEGGRG GSDGGGEGIQ DRESSFRFQP FESYDSRPCL PEHNPYRPSY SYDYEFDLGS DRNGSFGGQY SECRDPARER GSLDGFMRGR GQGRFQDRSN PGTFMRSDPF VPPAASSEPL STPWNELNYV GGRGLGGPSP SRPPPSLFSQ SMAPDYGVMG MQGAGGYDST MPYGCGRSQP RMRDRDRPKR RGFDRFGPDG TGRKRKQFQL YEEPDTKLAR VDSEGDFSEN DDAAGDFRSG DEEFKGEDEL CDSGRQRGEK EDEDEDVKKR REKQRRRDRT RDRAADRIQF ACSVCKFRSF DDEEIQKHLQ SKFHKETLRF ISTKLPDKTV EFLQEYIVNR NKKIEKRRQE LMEKETAKPK PDPFKGIGQE HFFKKIEAAH CLACDMLIPA QPQLLQRHLH SVDHNHNRRL AAEQFKKTSL HVAKSVLNNR HIVKMLEKYL KGEDPFTSET VDPEMEGDDN LGGEDKKETP EEVAADVLAE VITAAVRAVD GEGAPAPESS GEPAEDEGPT DTAEAGSDPQ AEQLLEEQVP CGTAHEKGVP KARSEAAEAG NGAETMAAEA ESAQTRVAPA PAAADAEVEQ TDAESKDAVP TE
The type II cAMP-protein kinase (PKA) is a multifunctional kinase with a broad range of substrates. Specificity of PKA signaling is thought to be mediated by the compartmentalization of the kinase to specific sites within the cell. To maintain this specific localization, the R subunit (RII) of PKA interacts with specific RII-anchoring proteins. The family of RII-anchoring proteins has been designated A-kinase anchoring proteins (AKAP). AKAP 95, also known as AKAP 8, is a nuclear matrix protein predominantly expressed in liver, heart, pancreas, kidney and skeletal muscle. During mitosis, AKAP 95 is recruited to the chromosomes and plays an essential role in mitotic progression. Characteristic of its family, AKAP 95 participates in PKA signaling through an interaction with the RII regulatory subunit. In addition, AKAP 95 forms a complex with HA95 and HDAC3 and is required for the deacetylation of Histone H3 in mitosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A kinase (PRKA) anchor protein 8; A-kinase anchor protein 8; A-kinase anchor protein 95 kDa; A-kinase anchor protein, 95kDa; AKAP 8; AKAP 95; AKAP-8; DKFZp586B1222
Gene Aliases: AKAP 95; AKAP-8; AKAP-95; AKAP8; AKAP95
UniProt ID: (Human) O43823
Entrez Gene ID: (Human) 10270
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support