Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: HSWEMPSSD TNANTSASGS ASADSVLSRI NQRLDMVPHL ETDMMQGGVY GSGGERYDSY ESCDSRAVLS ERDLYRSGYD YSELDPEMEM AYEGQYDAYR DQFRMRGNDT FGPRAQGWAR DARSGRPMAS GYGRMWEDPM GARGQCMSGA SRLPSLFSQN IIPEYGMFQG MRGGGAFPGG SRFGFGFGNG MKQMRRTWKT WTTADFRTKK KKRKQGGSPD E (67-286 aa encoded by BC000713)
Could play a role in constitutive transport element (CTE)-mediated gene expression by association with DHX9. Increases CTE-dependent nuclear unspliced mRNA export (PubMed:10748171, PubMed:11402034). Proposed to target PRKACA to the nucleus but does not seem to be implicated in the binding of regulatory subunit II of PKA (PubMed:10761695, PubMed:11884601). May be involved in nuclear envelope breakdown and chromatin condensation. May be involved in anchoring nuclear membranes to chromatin in interphase and in releasing membranes from chromating at mitosis (PubMed:11034899). May regulate the initiation phase of DNA replication when associated with TMPO isoform Beta (PubMed:12538639). Required for cell cycle G2/M transition and histone deacetylation during mitosis. In mitotic cells recruits HDAC3 to the vicinity of chromatin leading to deacetylation and subsequent phosphorylation at 'Ser-10' of histone H3; in this function seems to act redundantly with AKAP8 (PubMed:16980585). May be involved in regulation of pre-mRNA splicing (PubMed:17594903). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A kinase (PRKA) anchor protein 8-like; A-kinase anchor protein 8-like; AKAP8-like protein; HA95; HAP95; Helicase A-binding protein 95; helicase A-binding protein 95 kDa; Homologous to AKAP95 protein; neighbor of A-kinase anchoring protein 95; Neighbor of A-kinase-anchoring protein 95; neighbor of AKAP; Neighbor of AKAP95; testis tissue sperm-binding protein Li 90mP
Gene Aliases: AKAP8L; HA95; HAP95; HRIHFB2018; NAKAP; NAKAP95
UniProt ID: (Human) Q9ULX6
Entrez Gene ID: (Human) 26993
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support