Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MGDIPAVGLS SWKASPGKVT EAVKEAIDAG YRHFDCAYFY HNEREVGAGI RCKIKEGAVR REDLFIATKL WCTCHKKSLV ETACRKSLKA LKLNYLDLYL IHWPMGFKPP HPEWIMSCSE LSFCLSHPRV QDLPLDESNM VIPSDTDFLD TWEAMEDLVI TGLVKNIGVS NFNHEQLERL LNKPGLRFKP LTNQIECHPY LTQKNLISFC QSRDVSVTAY RPLGGSCEGV DLIDNPVIKR IAKEHGKSPA QILIRFQIQR NVIVIPGSIT PSHIKENIQV FDFELTQHDM DNILSLNRNL RLAMFPM
The protein encoded by this gene is a member of the aldo-keto reductase superfamily. Members in this family are characterized by their structure (evolutionarily highly conserved TIM barrel) and function (NAD(P)H-dependent oxido-reduction of carbonyl groups). Transcripts of this gene have been reported in specimens of human testis. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1,5-anhydro-D-fructose reductase; AF reductase; Aldo-keto reductase family 1 member C-like protein 2; Aldo-keto reductase family 1 member E2; aldo-keto reductase family 1, member C-like 2; aldo-keto reductase family 1, member E2; aldo-keto reductase loopADR; htAKR; hTSP; LoopADR; Testis aldo-keto reductase; Testis-specific protein
Gene Aliases: AKR1CL2; AKR1E2; AKRDC1; htAKR; hTSP; LoopADR
UniProt ID: (Human) Q96JD6
Entrez Gene ID: (Human) 83592
Molecular Function: oxidoreductase reductase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support