Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Recommended positive controls: 293T, A431, Jurkat, Raji.
Predicted reactivity: Rat (90%), Pig (91%), Rhesus Monkey (96%), Bovine (89%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
AKR7A2 is aldo-keto reductases, which are involved in the detoxification of aldehydes and ketones.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AFB1 aldehyde reductase 1; AFB1-AR 1; aflatoxin aldehyde reductase; Aflatoxin B1 aldehyde reductase member 2; aflatoxin beta1 aldehyde reductase; aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase); Aldoketoreductase 7; epididymis secretory sperm binding protein Li 166mP; HEL-S-166mP; SSA reductase; Succinic semialdehyde reductase; synonyms: AFAR1
Gene Aliases: AFAR; AFAR1; AFB1-AR1; AKR7; AKR7A2
UniProt ID: (Human) O43488
Entrez Gene ID: (Human) 8574
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support