Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLSQVYRCGF QPFNQHLLPW VKCTTVFRSH CIQPSVIRHV RSWSNIPFIT VPLSRTHGKS FAHRSELKHA KRIVVKLGSA VVTRGDECGL ALGRLASIVE QVSVLQNQGR EMMLVTSGAV AFGKQRLRHE ILLSQSVRQA LHSGQNQLKE MAIPVLEARA CAAAGQSGLM ALYEAMFTQY SICAAQILVT NLDFHDEQKR RNLNGTLHEL LRMNIVPIVN TNDAVVPPAE PNSDLQGVNV ISVKDNDSLA ARLAVEMKTD LLIVLSDVEG LFDSPPGSDD AKLIDIFYPG DQQSVTFGTK SRVGMGGMEA KVKAALWALQ GGTSVVIANG THPKVSGHVI TDIVEGKKVG TFFSEVKPAG PTVEQQGEMA RSGGRMLATL EPEQRAEIIH HLADLLTDQR DEILLANKKD LEEAEGRLAA PLLKRLSLST SKLNSLAIGL RQIAASSQDS VGRVLRRTRI AKNLELEQVT VPIGVLLVIF ESRPDCLPQV AALAIASGNG LLLKGGKEAA HSNRILHLLT QEALSIHGVK EAVQLVNTRE EVEDLCRLDK MIDLIIPRGS SQLVRDIQKA AKGIPVMGHS EGICHMYVDS EASVDKVTRL VRDSKCEYPA ACNALETLLI HRDLLRTPLF DQIIDMLRVE QVKIHAGPKF ASYLTFSPSE VKSLRTEYGD LELCIEVVDN VQDAIDHIHK YGSSHTDVIV TEDENTAEFF LQHVDSACVF WNASTRFSDG YRFGLGAEVG ISTSRIHARG PVGLEGLLTT KWLLRGKDHV VSDFSEHGSL KYLHENLPIP QRNTN
This gene is a member of the aldehyde dehydrogenase family and encodes a bifunctional ATP- and NADPH-dependent mitochondrial enzyme with both gamma-glutamyl kinase and gamma-glutamyl phosphate reductase activities. The encoded protein catalyzes the reduction of glutamate to delta1-pyrroline-5-carboxylate, a critical step in the de novo biosynthesis of proline, ornithine and arginine. Mutations in this gene lead to hyperammonemia, hypoornithinemia, hypocitrullinemia, hypoargininemia and hypoprolinemia and may be associated with neurodegeneration, cataracts and connective tissue diseases. Alternatively spliced transcript variants, encoding different isoforms, have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aldehyde dehydrogenase 18 family, member A1; Aldehyde dehydrogenase family 18 member A1; Delta-1-pyrroline-5-carboxylate synthase; delta-1-pyrroline-5-carboxylate synthetase; delta1-pyrroline-5-carboxlate synthetase; MGC117316; P5CS; pyrroline-5-carboxylate synthetase (glutamate gamma-semialdehyde synthetase)
Gene Aliases: ADCL3; ALDH18A1; ARCL3A; GSAS; P5CS; PYCS; SPG9A; SPG9B
UniProt ID: (Human) P54886
Entrez Gene ID: (Human) 5832
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support