Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Recommended positive controls: MCF-7, mouse testis, rat testis.
Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (81%), Japanese Medaka (81%), Xenopus laevis (88%), Dog (99%), Chicken (95%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This protein belongs to the aldehyde dehydrogenase family of proteins. The product of this gene is an enzyme that catalyzes the synthesis of retinoic acid (RA) from retinaldehyde. Retinoic acid, the active derivative of vitamin A (retinol), is a hormonal signaling molecule that functions in developing and adult tissues. The studies of a similar mouse gene suggest that this enzyme and the cytochrome CYP26A1, concurrently establish local embryonic retinoic acid levels which facilitate posterior organ development and prevent spina bifida. Three transcript variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AL1A2; alcohol dehydrogenase family 1, subfamily A2; alcohol dehydrogenase family 1, subfamily A7; aldehyde dehydrogenase 1 family, member A2; aldehyde dehydrogenase 1A2; Aldehyde dehydrogenase family 1 member A2; aldehyde dehydrogenase family 1, subfamily A2; ALDH1A2; RALDH 2; RALDH(II); ralDH2; Retinal dehydrogenase 2; retinal dehydrogenase, type II; retinaldehyde dehydrogenase 2; Retinaldehyde-specific dehydrogenase type 2
Gene Aliases: ALDH1A2; Aldh1a7; AV116159; RALDH(II); Raldh-2; Raldh1; RALDH2; RALDH2-T
UniProt ID: (Human) O94788, (Mouse) Q62148, (Rat) Q63639
Entrez Gene ID: (Human) 8854, (Mouse) 19378, (Rat) 116676
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support