Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MLRFLAPRLL SLQGRTARYS SAAALPSPIL NPDIPYNQLF INNEWQDAVS KKTFPTVNPT TGEVIGHVAE GDRADVDRAV KAAREAFRLG SPWRRMDASE RGRLLNLLAD LVERDRVYLA SLETLDNGKP FQESYALDLD EVIKVYRYFA GWADKWHGKT IPMDGQHFCF TRHEPVGVCG QIIPWNFPLV MQGWKLAPAL ATGNTVVMKV AEQTPLSALY LASLIKEAGF PPGVVNIITG YGPTAGAAIA QHMDVDKVAF TGSTEVGHLI QKAAGDSNLK RVTLELGGKS PSIVLADADM EHAVEQCHEA LFFNMGQCCC AGSRTFVEES IYNEFLERTV EKAKQRKVGN PFELDTQQGP QVDKEQFERV LGYIQLGQKE GAKLLCGGER FGERGFFIKP TVFGGVQDDM RIAKEEIFGP VQPLFKFKKI EEVVERANNT RYGLAAAVFT RDLDKAMYFT QALQAGTVWV NTYNIVTCHT PFGGFKESGN GRELGEDGLK AYTEVKTVTI KVPQKNS
Aldehyde dehydrogenases (ALDHs) mediate NADP+-dependent oxidation of aldehydes into acids during detoxification of alcohol-derived acetaldehyde, lipid peroxidation and metabolism of corticosteroids, biogenic amines and neurotransmitters. Alcohol drinking habits and cardiovascular disease risk factors may be associated with ALDH gene variants. ALDH1B1 (Aldehyde dehydrogenase family 1 member B1), also known as ALDH5 or ALDHX (Aldehyde dehydrogenase X, mitochondrial), is a 517 amino acid mitochondrial protein that is expressed in the liver, testis and to a lesser extent in brain. ALDH1B1 belongs to the aldehyde dehydrogenase family and may play a major role in ethanol detoxification.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetaldehyde dehydrogenase 5; aldehyde dehydrogenase 1 family, member B1; Aldehyde dehydrogenase 5; Aldehyde dehydrogenase family 1 member B1; Aldehyde dehydrogenase X, mitochondrial; ALDH class 2; MGC2230
Gene Aliases: ALDH1B1; ALDH5; ALDHX
UniProt ID: (Human) P30837
Entrez Gene ID: (Human) 219
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support