Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MELEVRRVR QAFLSGRSRP LRFRLQQLEA LRRMVQEREK DILTAIAADL CKSEFNVYSQ EVITVLGEID FMLENLPEWV TAKPVKKNVL TMLDEAYIQP QPLGVVLIIG AWNYPFVLTI QPLIGAIAAG NAVIIKPSEL SENTAKILAK LLPQYLDQDL YIVINGGVEE TTELLKQRFD HIFYTGNTAV GKIVMEAAAK HLTPVTLELG GKSPCYIDKD CDLDIVCRRI TWGKYMNCGQ TCIAPDYILC EASLQNQIVW KIKETVKEFY GENIKESPDY ERIINLRHFK RILSLLEGQK IAFGGETDEA TRYIA (1-314 aa encoded by BC002430)
Aldh3A2 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldehyde dehydrogenase enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. Aldh3A2 catalyzes the oxidation of long-chain aliphatic aldehydes to fatty acids. Mutations in the Aldh3A2 gene cause Sjogren-Larrson syndrome, an inherited neurocutaneous disorder. Patients with this disorder display ichthyosis, mental retardation and spastic diplegia. The pathogenesis of the cutaneous and neurological symptoms is thought to result from abnormal lipid accumulation in the membranes of skin and brain, the formation of aldehyde Schiff base adducts with amine-containing lipids or proteins, or defective eicosanoid metabolism.
仅用于科研。不用于诊断过程。未经明确授权不得转售。
蛋白别名: alcohol dehydrogenase family 3, subfamily A2; Aldehyde dehydrogenase 10; Aldehyde dehydrogenase 3; aldehyde dehydrogenase 3 family, member A2; aldehyde dehydrogenase 3A2; Aldehyde dehydrogenase 4; Aldehyde dehydrogenase family 3 member A2; aldehyde dehydrogenase family 3, subfamily A2; DKFZp686E23276; Fatty aldehyde dehydrogenase; FLJ20851; Microsomal aldehyde dehydrogenase; msALDH
基因别名: Ahd-3; Ahd-3r; Ahd3; Ahd3-r; AI194803; ALDH10; Aldh3; ALDH3A2; Aldh4; Aldh4-r; FALDH; SLS
UniProt ID: (Human) P51648, (Rat) P30839, (Mouse) P47740
Entrez Gene ID: (Human) 224, (Rat) 65183, (Mouse) 11671