Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with bovine, canine and porcine based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This protein belongs to the aldehyde dehydrogenases family of proteins. This enzyme plays a role in the valine and pyrimidine catabolic pathways. The product of this gene, a mitochondrial methylmalonate semialdehyde dehydrogenase, catalyzes the irreversible oxidative decarboxylation of malonate and methylmalonate semialdehydes to acetyl- and propionyl-CoA. Methylmalonate semialdehyde dehydrogenase deficiency is characterized by elevated beta-alanine, 3-hydroxypropionic acid, and both isomers of 3-amino and 3-hydroxyisobutyric acids in urine organic acids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aldehyde dehydrogenase 6 family, member A1; Aldehyde dehydrogenase family 6 member A1; malonate-semialdehyde dehydrogenase; Malonate-semialdehyde dehydrogenase [acylating]; methylmalonate-semialdehyde dehydrogenase [acylating], mitochondrial; Methylmalonate-semialdehyde/malonate-semialdehyde dehydrogenase [acylating], mitochondrial; MGC40271; mitochondrial acylating methylmalonate-semialdehyde dehydrogenase; MMSDH; testicular tissue protein Li 122
Gene Aliases: 1110038I05Rik; AI314632; ALDH6A1; MMSADHA; MMSDH
UniProt ID: (Human) Q02252, (Mouse) Q9EQ20
Entrez Gene ID: (Human) 4329, (Mouse) 104776
Molecular Function: dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support